accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CSCl

CAS RN 635305-24-7

IUPAC Name: 2-(5-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(5-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 244.54
Molecular Formula: C10H14BClO2S
InChi Key: DDDRRTOIHWNUSI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(Cl)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results