missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentyne, 98%

Catalog No. AC223420050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223420050 5 g Glass bottle
AC223420250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223420050 Supplier Thermo Scientific Chemicals Supplier No. 223420050

CAS: 14267-92-6 | C5H7Cl | 102.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14267-92-6
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.56
MDL Number MFCD00001014
InChI Key UXFIKVWAAMKFQE-UHFFFAOYSA-N
Synonym 5-chloro-1-pentyne, 1-pentyne, 5-chloro, 5-chloropentyne, 4-pentynyl chloride, 1-chloro-4-pentine, 1-chloro-4-pentyne, pent-4-ynyl chloride, 5-chloro-pent-1-yne, acmc-1bzfd, pent-4-yn-1-yl chloride
PubChem CID 84308
IUPAC Name 5-chloropent-1-yne
SMILES C#CCCCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Brown
Density 0.9600g/mL
Boiling Point 67°C to 69°C (145.0 mmHg)
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4430 to 1.445
Linear Formula Cl(CH2)3C∽CH
Quantity 5 g
Beilstein 01, III, 958
Specific Gravity 0.96
Formula Weight 102.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Chloro-1-pentyne, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-159-5
TSCA TSCA

RUO – Research Use Only