Learn More
5-Chloro-1-pentanol, 95%
CAS: 5259-98-3 | C5H11ClO | 122.59 g/mol
Supplier: Thermo Scientific Chemicals L1208818
Description
5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 5-Chloro-1-pentanol | |
| 1.06 | |
| C5H11ClO | |
| MFCD00039556 | |
| 1732314 | |
| Slightly soluble in water. | |
| OCCCCCCl | |
| 122.59 | |
| 122.6 |
| 5259-98-3 | |
| 102°C to 103°C (8 mmHg) | |
| 1.454 | |
| 50 g | |
| 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l | |
| DCBJCKDOZLTTDW-UHFFFAOYSA-N | |
| 5-chloropentan-1-ol | |
| 78915 | |
| 95% |
Chemical Identifiers
| 5259-98-3 | |
| 122.59 | |
| DCBJCKDOZLTTDW-UHFFFAOYSA-N | |
| 78915 | |
| OCCCCCCl |
| C5H11ClO | |
| MFCD00039556 | |
| 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l | |
| 5-chloropentan-1-ol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 226-067-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only