missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentanol, 95%

Catalog No. AAL1208809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1208809 10 g
AAL1208818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1208809 Supplier Thermo Scientific Chemicals Supplier No. L1208809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5259-98-3 | C5H11ClO | 122.59 g/mol

5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5259-98-3
Molecular Formula C5H11ClO
Molecular Weight (g/mol) 122.59
MDL Number MFCD00039556
InChI Key DCBJCKDOZLTTDW-UHFFFAOYSA-N
Synonym 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l
PubChem CID 78915
IUPAC Name 5-chloropentan-1-ol
SMILES OCCCCCCl

Specifications

Density 1.06
Boiling Point 102°C to 103°C (8 mmHg)
Refractive Index 1.454
Quantity 10 g
Beilstein 1732314
Solubility Information Slightly soluble in water.
Formula Weight 122.6
Percent Purity 95%
Chemical Name or Material 5-Chloro-1-pentanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-067-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only