missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methylisatin 98.0+%, TCI America™

Catalog No. p-7125885
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3195200MG 200 mg
C31951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3195200MG Supplier TCI America Supplier No. C3195200MG

Chemical Identifiers

CAS 60434-13-1
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.60
MDL Number MFCD02956479
InChI Key NJOPQQPDPYWFFA-UHFFFAOYSA-N
Synonym 5-Chloro-1-methylindole-2,3-dione, 5-Chloro-1-methyl-2,3-indolinedione
PubChem CID 108941
IUPAC Name 5-chloro-1-methyl-2,3-dihydro-1H-indole-2,3-dione
SMILES CN1C(=O)C(=O)C2=CC(Cl)=CC=C12

Specifications

Melting Point 177°C
Quantity 200 mg
Formula Weight 195.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1-methylisatin