accessibility menu, dialog, popup

UserName

H3CNOOCl

CAS RN 60434-13-1

IUPAC Name: 5-chloro-1-methyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-chloro-1-methylindole-2,3-dione
Molecular Weight (g/mol): 195.6
Molecular Formula: C9H6ClNO2
InChi Key: NJOPQQPDPYWFFA-UHFFFAOYSA-N
SMILES: CN1C(=O)C(=O)C2=CC(Cl)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results