missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,10-phenanthroline

Catalog No. p-7049917
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3118002 0.5 g
AA3118004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3118002 Supplier Thermo Scientific Chemicals Supplier No. 03118002
Only null left

CAS: 4199-89-7 | C12H7ClN2 | 214.65 g/mol

Used to prepare 1,10-phenanthrolinium cation. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4199-89-7
Molecular Formula C12H7ClN2
Molecular Weight (g/mol) 214.65
MDL Number MFCD00004980
InChI Key XDUUQOQFSWSZSM-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 5-chloro, 5-chloropyridino 3,2-h quinoline, acmc-209jmo, 1,10-phenanthroline,5-chloro, 5-chloro-1,10 phenanthroline, 5-chloro-1,10-phenanthroline, 5-chloro-1, 10-phenanthroline
PubChem CID 77865
IUPAC Name 5-chloro-1,10-phenanthroline
SMILES ClC1=CC2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 123°C
Quantity 0.5 g
Sensitivity Hygroscopic
Solubility Information It is soluble in organic solvents.
Formula Weight 214.66
Physical Form Crystalline
Chemical Name or Material 5-Chloro-1,10-phenanthroline
EINECSNumber 224-098-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only