missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,10-phenanthroline 98.0+%, TCI America™

Catalog No. p-7124606
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0786100MG 100 mg
C07861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0786100MG Supplier TCI America Supplier No. C0786100MG
Only null left

Chemical Identifiers

CAS 4199-89-7
Molecular Formula C12H7ClN2
Molecular Weight (g/mol) 214.65
MDL Number MFCD00004980
InChI Key XDUUQOQFSWSZSM-UHFFFAOYSA-N
PubChem CID 77865
IUPAC Name 5-chloro-1,10-phenanthroline
SMILES ClC1=CC2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 123°C
Quantity 100 mg
Synonym 1,10-phenanthroline, 5-chloro, 5-chloropyridino 3,2-h quinoline, acmc-209jmo, 1,10-phenanthroline,5-chloro, 5-chloro-1,10 phenanthroline, 5-chloro-1,10-phenanthroline, 5-chloro-1, 10-phenanthroline
Formula Weight 214.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1,10-phenanthroline