missing translation for 'onlineSavingsMsg'
Learn More

(5-Carboxypentyl)triphenylphosphonium Bromide 96.0+%, TCI America™

Catalog No. C31135G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31135G Supplier TCI America Supplier No. C31135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50889-29-7
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00055556
InChI Key JUWYRPZTZSWLCY-UHFFFAOYSA-N
Synonym 5-carboxypentyl triphenylphosphonium bromide, 5-carboxypentyl triphenyl phosphonium bromide, 5-carboxypentyl triphenylphosphanium bromide, 5-carboxypentyltriphenylphosphonium bromide, phosphonium, 5-carboxypentyl triphenyl-, bromide, acmc-20apmi, pxjvnuuwrjumaq-uhfffaoysa-m, 5-carboxyamyl triphenylphosphonium bromide
PubChem CID 2779280
IUPAC Name (5-carboxypentyl)triphenylphosphanium bromide
SMILES [Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 5 g
Formula Weight 457.35
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (5-Carboxypentyl)triphenylphosphonium Bromide