missing translation for 'onlineSavingsMsg'
Learn More

(5-Carboxypentyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog No. AAB2058806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2058806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2058806 Supplier Thermo Scientific Chemicals Supplier No. B2058806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50889-29-7 | Molecular Formula: C24H26BrO2P | Molecular Weight: 457.35 g/mol

It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.

Solubility
Soluble in Chloroform and Methanol.

Notes
Hygroscopic. Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 50889-29-7
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00055556
InChI Key JUWYRPZTZSWLCY-UHFFFAOYSA-N
Synonym 5-carboxypentyl triphenylphosphonium bromide, 5-carboxypentyl triphenyl phosphonium bromide, 5-carboxypentyl triphenylphosphanium bromide, 5-carboxypentyltriphenylphosphonium bromide, phosphonium, 5-carboxypentyl triphenyl-, bromide, acmc-20apmi, pxjvnuuwrjumaq-uhfffaoysa-m, 5-carboxyamyl triphenylphosphonium bromide
PubChem CID 2779280
IUPAC Name 5-carboxypentyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 200°C to 202°C
Quantity 5 g
Solubility Information Soluble in Chloroform and Methanol.
Formula Weight 457.36
Percent Purity 97%
Chemical Name or Material (5-Carboxypentyl)triphenylphosphonium bromide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only