missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxyfluorescein Diacetate 93.0+%, TCI America™

Catalog No. p-7125687
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C285950MG 50 mg
C2859200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C285950MG Supplier TCI America Supplier No. C285950MG
Only null left

Chemical Identifiers

CAS 79955-27-4
Molecular Formula C25H16O9
Molecular Weight (g/mol) 460.394
MDL Number MFCD00036872
InChI Key WPUZGNPQMIWOHE-UHFFFAOYSA-N
Synonym 5-carboxyfluorescein diacetate, 5-cfda, 3',6'-diacetoxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid, 5-carboxy-di-o-acetylfluorescein, 3',6'-diacetyloxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, cfda-5, spiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylicacid, 3',6'-bis acetyloxy-3-oxo, 3',6'-bis acetyloxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-bis acetyloxy-3-oxospiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylic acid, 3',6'-diacetoxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid
PubChem CID 133314
IUPAC Name 3',6'-diacetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(C=C(C=C4)C(=O)O)C(=O)O3)C5=C(O2)C=C(C=C5)OC(=O)C

Specifications

Quantity 50 mg
Formula Weight 460.39
Percent Purity ≥93.0% (HPLC,T)
Chemical Name or Material 5-Carboxyfluorescein Diacetate
TSCA No
Recommended Storage Freezer