missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
$596.67 - $596.67
Chemical Identifiers
| CAS | 1412905-42-0 |
|---|---|
| Molecular Formula | C13H17BFNO3 |
| Molecular Weight (g/mol) | 265.09 |
| MDL Number | MFCD18730404 |
| InChI Key | RBEAFMIZDXOHAT-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester |
| PubChem CID | 99738340 |
| IUPAC Name | 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide |
| SMILES | CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5926203
|
Thermo Scientific Chemicals
H5926203 |
1 g |
Each for $596.67
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1412905-42-0 | |
| 265.09 | |
| RBEAFMIZDXOHAT-UHFFFAOYSA-N | |
| 99738340 | |
| CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O |
| C13H17BFNO3 | |
| MFCD18730404 | |
| 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester | |
| 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide |
Specifications
| 1412905-42-0 | |
| MFCD18730404 | |
| 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester | |
| CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O | |
| 265.09 | |
| 265.09 | |
| 5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester |
| C13H17BFNO3 | |
| 1 g | |
| RBEAFMIZDXOHAT-UHFFFAOYSA-N | |
| 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide | |
| 99738340 | |
| 96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures