missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH5926203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5926203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5926203 Supplier Thermo Scientific Chemicals Supplier No. H5926203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1412905-42-0
Molecular Formula C13H17BFNO3
Molecular Weight (g/mol) 265.09
MDL Number MFCD18730404
InChI Key RBEAFMIZDXOHAT-UHFFFAOYSA-N
Synonym 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 99738340
IUPAC Name 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O

Specifications

Quantity 1 g
Formula Weight 265.09
Percent Purity 96%
Chemical Name or Material 5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures