missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoyl-2-fluorobenzeneboronic acid, 95%

Catalog No. AAH53010MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53010MD 250 mg
AAH5301003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53010MD Supplier Thermo Scientific Chemicals Supplier No. H53010MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 874289-39-1 | C7H7BFNO3 | 182.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874289-39-1
Molecular Formula C7H7BFNO3
Molecular Weight (g/mol) 182.95
MDL Number MFCD08436055
InChI Key NDVPMVJEURRBQM-UHFFFAOYSA-N
Synonym 3-borono-4-fluorobenzamide, 5-carbamoyl-2-fluorophenyl boronic acid, 5-carbamoyl-2-fluorobenzeneboronic acid, 5-carbamoyl-2-fluorophenyboronic acid, acmc-209qmh, 2-fluoro-5-carbamoylphenylboronic acid, 5-carbamoyl-2-fluorophenyl boronicacid, 5-carbamoyl-2-fluoro-phenyl boronic acid, 5-carbamoyl-2-fluorobenzeneboronicacid
PubChem CID 44717478
IUPAC Name (5-carbamoyl-2-fluorophenyl)boronic acid
SMILES NC(=O)C1=CC(B(O)O)=C(F)C=C1

Specifications

Melting Point 192°C to 194°C
Quantity 250 mg
Formula Weight 182.95
Percent Purity 95%
Chemical Name or Material 5-Carbamoyl-2-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only