missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Butylpyridine-2-carboxylic Acid 98.0+%, TCI America™
$710.21 - $710.21
Chemical Identifiers
| CAS | 536-69-6 |
|---|---|
| Molecular Formula | C10H13NO2 |
| Molecular Weight (g/mol) | 179.219 |
| MDL Number | MFCD00006298 |
| InChI Key | DGMPVYSXXIOGJY-UHFFFAOYSA-N |
| Synonym | fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acid |
| PubChem CID | 3442 |
| ChEBI | CHEBI:5199 |
| IUPAC Name | 5-butylpyridine-2-carboxylic acid |
| SMILES | CCCCC1=CN=C(C=C1)C(=O)O |
Chemical Identifiers
| 536-69-6 | |
| 179.219 | |
| DGMPVYSXXIOGJY-UHFFFAOYSA-N | |
| 3442 | |
| 5-butylpyridine-2-carboxylic acid |
| C10H13NO2 | |
| MFCD00006298 | |
| fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acid | |
| CHEBI:5199 | |
| CCCCC1=CN=C(C=C1)C(=O)O |
Specifications
| 536-69-6 | |
| White-Yellow | |
| MFCD00006298 | |
| fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acid | |
| CCCCC1=CN=C(C=C1)C(=O)O | |
| 179.219 | |
| CHEBI:5199 | |
| ≥98.0% (GC,T) | |
| 5-Butylpyridine-2-carboxylic Acid |
| 98°C | |
| C10H13NO2 | |
| 1 g | |
| DGMPVYSXXIOGJY-UHFFFAOYSA-N | |
| 5-butylpyridine-2-carboxylic acid | |
| 3442 | |
| 179.22 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : US5625000
TSCA : Yes
Recommended Storage : Refrigerator