accessibility menu, dialog, popup

UserName

H3CNOHO

CAS RN 536-69-6

IUPAC Name: 5-butylpyridine-2-carboxylic acid
Synonyms: fusaric acid 5 butyl 2 pyridinedicarboxylic acid acid, fusaric
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: DGMPVYSXXIOGJY-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(N=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)