missing translation for 'onlineSavingsMsg'
Learn More

5-Butylpyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. F02271G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02271G Supplier TCI America Supplier No. F02271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 536-69-6
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00006298
InChI Key DGMPVYSXXIOGJY-UHFFFAOYSA-N
Synonym fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acid
PubChem CID 3442
ChEBI CHEBI:5199
IUPAC Name 5-butylpyridine-2-carboxylic acid
SMILES CCCCC1=CN=C(C=C1)C(=O)O

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Butylpyridine-2-carboxylic Acid
RTECSNumber US5625000
TSCA Yes
Recommended Storage Refrigerator