missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleryl Chloride 98.0+%, TCI America™

Catalog No. B20595G
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20595G 5 g
B205925G 25 g
B2059250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B20595G Supplier TCI America Supplier No. B20595G

Chemical Identifiers

CAS 4509-90-4
Molecular Formula C14H11N3O2
Molecular Weight (g/mol) 253.26
MDL Number MFCD00013660
InChI Key QANQQXSQVQYQFI-UHFFFAOYSA-N
Synonym 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride
PubChem CID 521173
IUPAC Name N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide
SMILES CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 3265
Formula Weight 199.47
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 5-Bromovaleryl Chloride