missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleryl chloride, 98%

Catalog No. AAA1804606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1804606 5 g
AAA1804614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1804606 Supplier Thermo Scientific Chemicals Supplier No. A1804606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4509-90-4 | C14H11N3O2 | 253.26 g/mol

5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.

Chemical Identifiers

CAS 4509-90-4
Molecular Formula C14H11N3O2
Molecular Weight (g/mol) 253.26
MDL Number MFCD00013660
InChI Key QANQQXSQVQYQFI-UHFFFAOYSA-N
Synonym 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride
PubChem CID 521173
IUPAC Name 5-bromopentanoyl chloride
SMILES CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1

Specifications

Density 1.49
Boiling Point 43°C to 45°C (0.2 mmHg)
Flash Point 107°C (225°F)
Odor Irritating
Refractive Index 1.491
Linear Formula Br(CH2)4COCl
Quantity 5 g
UN Number UN3265
Beilstein 1745663
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 199.46
Percent Purity 98%
Chemical Name or Material 5-Bromovaleryl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only