missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleronitrile 97.0+%, TCI America™

Catalog No. p-7119060
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B14535G 5 g
B145325G 25 g
2 options

Catalog No. B14535G

Supplier: TCI America B14535G

Only null left

Chemical Identifiers

CAS 5414-21-1
Molecular Formula C5H8BrN
Molecular Weight (g/mol) 162.03
MDL Number MFCD00001976
InChI Key NWWWGAKVHCSAEU-UHFFFAOYSA-N
Synonym 5-bromovaleronitrile, pentanenitrile, 5-bromo, 5bromovaleronitrile, snxhflqiuzjp@, 5-bromopentanonitrile, 4-bromobutyl cyanide, 5-bromo-valeronitrile, 5-bromo-pentanenitrile, 5-bromopentane nitrile, 1-bromo-4-cyanobutane
PubChem CID 79435
IUPAC Name 5-bromopentanenitrile
SMILES C(CCBr)CC#N

Specifications

Color Yellow
Boiling Point 111°C
Quantity 5 g
UN Number 3276
Formula Weight 162.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromovaleronitrile
EINECSNumber (2)-3503
TSCA No