missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleronitrile, 98+%

Catalog No. AAL0395214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0395214 25 g
AAL0395206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0395214 Supplier Thermo Scientific Chemicals Supplier No. L0395214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5414-21-1 | C5H8BrN | 162.03 g/mol

5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

Solubility
Slightly soluble in water (4.1 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5414-21-1
Molecular Formula C5H8BrN
Molecular Weight (g/mol) 162.03
MDL Number MFCD00001976
InChI Key NWWWGAKVHCSAEU-UHFFFAOYSA-N
Synonym 5-bromovaleronitrile, pentanenitrile, 5-bromo, 5bromovaleronitrile, snxhflqiuzjp@, 5-bromopentanonitrile, 4-bromobutyl cyanide, 5-bromo-valeronitrile, 5-bromo-pentanenitrile, 5-bromopentane nitrile, 1-bromo-4-cyanobutane
PubChem CID 79435
IUPAC Name 5-bromopentanenitrile
SMILES C(CCBr)CC#N

Specifications

Density 1.388
Boiling Point 110°C to 111°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4785
Quantity 25 g
UN Number UN3276
Beilstein 1742080
Solubility Information Slightly soluble in water (4.1g/L at 25°C).
Formula Weight 162.04
Percent Purity ≥98%
Chemical Name or Material 5-Bromovaleronitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 226-505-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only