missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxylic acid hydrazide, 97%

Catalog No. p-7041395
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6195906 5 g
AAH6195914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6195906 Supplier Thermo Scientific Chemicals Supplier No. H6195906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98027-27-1 | C5H5BrN2OS | 221.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98027-27-1
Molecular Formula C5H5BrN2OS
Molecular Weight (g/mol) 221.07
MDL Number MFCD01310798
InChI Key OFXZACLPQYOIAV-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxylic acid hydrazide, 5-bromo-thiophene-2-carboxylic acid hydrazide, 5-bromo-thiophene-2-carboxylic acidhydrazide, 5-bromothiophene-2-carboxylic acid hydrazide, 2-thiophenecarboxylic acid, 5-bromo-, hydrazide, pubchem16117, ksc488m6p, 5-bromo-2-thiophenecarboxylic hydrazide, 2-bromothiophene-5-carbohydrazide, 2-thiophenecarboxylicacid, 5-bromo-, hydrazide
PubChem CID 2063863
IUPAC Name 5-bromothiophene-2-carbohydrazide
SMILES NNC(=O)C1=CC=C(Br)S1

Specifications

Quantity 5 g
Formula Weight 221.07
Percent Purity 98%
Chemical Name or Material 5-Bromothiophene-2-carboxylic acid hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only