accessibility menu, dialog, popup

UserName

NH2HNOSBr

CAS RN 98027-27-1

IUPAC Name: 5-bromothiophene-2-carbohydrazide
Synonyms: 5-bromothiophene-2-carbohydrazide
Molecular Weight (g/mol): 221.07
Molecular Formula: C5H5BrN2OS
InChi Key: OFXZACLPQYOIAV-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results