missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxylic acid, 98%

Catalog No. AAA1924203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1924203 1 g
AAA1924206 5 g
AAA1924214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1924203 Supplier Thermo Scientific Chemicals Supplier No. A1924203

CAS: 7311-63-9 | C5H2BrO2S | 206.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7311-63-9
Molecular Formula C5H2BrO2S
Molecular Weight (g/mol) 206.03
MDL Number MFCD00079725
InChI Key COWZPSUDTMGBAT-UHFFFAOYSA-M
Synonym 5-bromo-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-bromo, 2-bromo-5-carboxythiophene, 2-bromothiophene-5-carboxylic acid, 5-bromo-thiophene-2-carboxylic acid, 5-bromothiophene-2-carboxylicacid, zlchem 973, pubchem7368, acmc-209opm, maybridge1_002056
PubChem CID 349115
IUPAC Name 5-bromothiophene-2-carboxylic acid
SMILES [O-]C(=O)C1=CC=C(Br)S1

Specifications

Melting Point 139°C to 143°C
Quantity 1 g
Beilstein 118364
Formula Weight 207.05
Percent Purity 98%
Chemical Name or Material 5-Bromothiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only