accessibility menu, dialog, popup

UserName

CAS RN 7311-63-9

IUPAC Name: 5-bromothiophene-2-carboxylate
Synonyms: 5-bromothiophene-2-carboxylate
Molecular Weight (g/mol): 206.03
Molecular Formula: C5H2BrO2S
InChi Key: COWZPSUDTMGBAT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(Br)S1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)