missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxaldehyde 97.0+%, TCI America™

Catalog No. p-7118871
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11355G 5 g
B113525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11355G Supplier TCI America Supplier No. B11355G
Only null left

Chemical Identifiers

CAS 4701-17-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005432
InChI Key GFBVUFQNHLUCPX-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
PubChem CID 78428
IUPAC Name 5-bromothiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C=O

Specifications

Boiling Point 115°C
Quantity 5 g
Formula Weight 191.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromothiophene-2-carboxaldehyde
TSCA Yes
Recommended Storage Refrigerator