missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxaldehyde, 97%

Catalog No. AAA1338106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1338106 5 g
AAA1338114 25 g
AAA1338122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1338106 Supplier Thermo Scientific Chemicals Supplier No. A1338106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4701-17-1 | C5H3BrOS | 191.04 g/mol

5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.

Solubility
Soluble in chloroform and methanol.

Notes
Air Sensitive, avoid contact with air/oxygen. Store in cool, dry place in tightly closed container.

Chemical Identifiers

CAS 4701-17-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005432
InChI Key GFBVUFQNHLUCPX-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
PubChem CID 78428
IUPAC Name 5-bromothiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C=O

Specifications

Density 1.762
Boiling Point 104°C to 106°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.639
Quantity 5 g
Beilstein 108404
Sensitivity Air sensitive
Solubility Information Soluble in chloroform and methanol.
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 5-Bromothiophene-2-carboxaldehyde
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-176-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only