missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoquinoline 98.0+%, TCI America™
Supplier: TCI America B30781G
Specifications
| 5-Bromoquinoline | |
| 46°C | |
| 280°C | |
| MFCD00234481 | |
| 5-bromo-quinoline, quinoline, 5-bromo, 5-bromooquinoline, 5-bromo quinoline, 5-bromoquinolline, pubchem7116, 5-bromoquinoline, acmc-209kh7, ksc492i0h, 5-bromoquinoline 250mg | |
| C1=CC2=C(C=CC=N2)C(=C1)Br | |
| 208.058 | |
| 208.06 | |
| Crystalline Powder |
| 4964-71-0 | |
| Green-Yellow | |
| C9H6BrN | |
| 1 g | |
| CHODTZCXWXCALP-UHFFFAOYSA-N | |
| 5-bromoquinoline | |
| 817321 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4964-71-0 | |
| 208.058 | |
| CHODTZCXWXCALP-UHFFFAOYSA-N | |
| 817321 | |
| C1=CC2=C(C=CC=N2)C(=C1)Br |
| C9H6BrN | |
| MFCD00234481 | |
| 5-bromo-quinoline, quinoline, 5-bromo, 5-bromooquinoline, 5-bromo quinoline, 5-bromoquinolline, pubchem7116, 5-bromoquinoline, acmc-209kh7, ksc492i0h, 5-bromoquinoline 250mg | |
| 5-bromoquinoline |
Safety and Handling
TSCA : No