accessibility menu, dialog, popup

UserName

BrN

CAS RN 4964-71-0

IUPAC Name: 5-bromoquinoline
Synonyms: 5-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: CHODTZCXWXCALP-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=NC2=CC=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)