missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoquinoline, 97%, Thermo Scientific Chemicals

Catalog No. p-7047691
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1963403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1963403 Supplier Thermo Scientific Chemicals Supplier No. L1963403
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4964-71-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00234481
InChI Key CHODTZCXWXCALP-UHFFFAOYSA-N
Synonym 5-bromo-quinoline, quinoline, 5-bromo, 5-bromooquinoline, 5-bromo quinoline, 5-bromoquinolline, pubchem7116, 5-bromoquinoline, acmc-209kh7, ksc492i0h, 5-bromoquinoline 250mg
PubChem CID 817321
IUPAC Name 5-bromoquinoline
SMILES C1=CC2=C(C=CC=N2)C(=C1)Br

Specifications

Melting Point 44°C to 48°C
Boiling Point 105°C to 107°C (1 mmHg)
Quantity 1 g
Beilstein 115148
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 5-Bromoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only