Learn More
5-Bromophthalide, 98%
CAS: 64169-34-2 | C8H5BrO2 | 213.03 g/mol
$63.55 - $628.45
Chemical Identifiers
| CAS | 64169-34-2 |
|---|---|
| Molecular Formula | C8H5BrO2 |
| Molecular Weight (g/mol) | 213.03 |
| MDL Number | MFCD01797360 |
| InChI Key | IUSPXLCLQIZFHL-UHFFFAOYSA-N |
| Synonym | 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide |
| PubChem CID | 603144 |
| IUPAC Name | 5-bromo-1,3-dihydro-2-benzofuran-1-one |
| SMILES | BrC1=CC=C2C(=O)OCC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3389106
|
Thermo Scientific Chemicals
H3389106 |
5 g |
Each for $63.55
|
|
|||||
|
AAH3389114
|
Thermo Scientific Chemicals
H3389114 |
25 g |
Each for $192.42
|
|
|||||
|
AAH3389122
|
Thermo Scientific Chemicals
H3389122 |
100 g |
Each for $628.45
|
|
|||||
Description
5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.
Solubility
Soluble in acetone.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 64169-34-2 | |
| 213.03 | |
| IUSPXLCLQIZFHL-UHFFFAOYSA-N | |
| 603144 | |
| BrC1=CC=C2C(=O)OCC2=C1 |
| C8H5BrO2 | |
| MFCD01797360 | |
| 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide | |
| 5-bromo-1,3-dihydro-2-benzofuran-1-one |
Specifications
| 64169-34-2 | |
| C8H5BrO2 | |
| 5 g | |
| Soluble in acetone. | |
| BrC1=CC=C2C(=O)OCC2=C1 | |
| 213.03 | |
| 213.03 | |
| 5-Bromophthalide |
| 162°C to 166°C | |
| MFCD01797360 | |
| 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide | |
| IUSPXLCLQIZFHL-UHFFFAOYSA-N | |
| 5-bromo-1,3-dihydro-2-benzofuran-1-one | |
| 603144 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 447-190-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only