missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromopentyl Acetate 95.0+%, TCI America™
Supplier: TCI America A11955G
Specifications
| 5-Bromopentyl Acetate | |
| Yellow | |
| C7H13BrO2 | |
| 5 g | |
| JIHJLQWUVZUKCH-UHFFFAOYSA-N | |
| 5-bromopentyl acetate | |
| 85140 | |
| ≥95.0% (GC) |
| 15848-22-3 | |
| 113°C | |
| MFCD00039204 | |
| 5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester | |
| CC(=O)OCCCCCBr | |
| 209.08 | |
| 209.08 | |
| Liquid |
Chemical Identifiers
| 15848-22-3 | |
| 209.08 | |
| JIHJLQWUVZUKCH-UHFFFAOYSA-N | |
| 85140 | |
| CC(=O)OCCCCCBr |
| C7H13BrO2 | |
| MFCD00039204 | |
| 5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester | |
| 5-bromopentyl acetate |
Safety and Handling
TSCA : No