missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopentyl acetate, 98%

Catalog No. p-7022666
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1554906 5 g
AAA1554914 25 g
AAA1554922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1554906 Supplier Thermo Scientific Chemicals Supplier No. A1554906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15848-22-3 | C7H13BrO2 | 209.08 g/mol

5-?Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-​Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 15848-22-3
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00039204
InChI Key JIHJLQWUVZUKCH-UHFFFAOYSA-N
Synonym 5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester
PubChem CID 85140
IUPAC Name 5-bromopentyl acetate
SMILES CC(=O)OCCCCCBr

Specifications

Density 1.255
Boiling Point 109°C to 110°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.462
Quantity 5 g
Beilstein 1752109
Solubility Information Slightly soluble in water.
Formula Weight 209.09
Percent Purity 98%
Chemical Name or Material 5-Bromopentyl acetate
EINECSNumber 239-961-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only