missing translation for 'onlineSavingsMsg'
Learn More

5-Bromonicotinic acid, 98%

Catalog No. p-3777494
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211390100 10 g Glass bottle
AC211390500 50 g Glass bottle
AC211392500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211390100 Supplier Thermo Scientific Chemicals Supplier No. 211390100

CAS: 20826-04-4 | C6H4BrNO2 | 202.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20826-04-4
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00009783
InChI Key FQIUCPGDKPXSLL-UHFFFAOYSA-N
Synonym 5-bromonicotinic acid, 5-bromo nicotinic acid, 5-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo, nicotinic acid, 5-bromo, 5-bromo-nicotinic acid, 3-bromo-5-pyridine carboxylic acid, 3-bromo pyridine-5-carboxylic acid, pubchem1211, acmc-209fev
PubChem CID 88707
IUPAC Name 5-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 178°C to 180°C
Color Brown or Gray-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula Br(C5H3N)COOH
Quantity 10 g
Beilstein 22, 44
Formula Weight 202.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromonicotinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-065-5

RUO – Research Use Only