accessibility menu, dialog, popup

UserName

CAS RN 20826-04-4

IUPAC Name: 5-bromopyridine-3-carboxylic acid
Synonyms: 5-bromonicotinic acid
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: FQIUCPGDKPXSLL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC(Br)=C1
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)