missing translation for 'onlineSavingsMsg'
Learn More

5-(bromomethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole, 97%, Thermo Scientific™

Catalog No. p-3880760
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC76308CB 250mg Amber Glass Bottle
CC76308DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC76308CB Supplier Thermo Scientific Chemicals Supplier No. CC76308CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1000339-73-0
Molecular Formula C6H5BrF3NS
Molecular Weight (g/mol) 260.07
MDL Number MFCD09264554
InChI Key OYDQGPCAYKWSAU-UHFFFAOYSA-N
Synonym 5-bromomethyl-2-methyl-4-trifluoromethyl-1,3-thiazole, 5-bromomethyl-2-methyl-4-trifluoromethyl thiazole, 5-bromomethyl-2-methyl-4-trifluoromethylthiazole, 5-bromomethyl-2-methyl-4-trifluoromethyl-thiazole
PubChem CID 26597955
IUPAC Name 5-(bromomethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole
SMILES CC1=NC(=C(CBr)S1)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 260.08
Percent Purity 97%
Chemical Name or Material 5-(Bromomethyl)-2-methyl-4-(trifluoromethyl)-1, 3-thiazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.