accessibility menu, dialog, popup

UserName

CH3NSBrFFF

CAS RN 1000339-73-0

IUPAC Name: 5-(bromomethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole
Synonyms: 5-(bromomethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole
Molecular Weight (g/mol): 260.07
Molecular Formula: C6H5BrF3NS
InChi Key: OYDQGPCAYKWSAU-UHFFFAOYSA-N
SMILES: CC1=NC(=C(CBr)S1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)