missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisatin, 98%

Catalog No. AC154230050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154230050 5 g Glass bottle
AC154230250 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154230050 Supplier Thermo Scientific Chemicals Supplier No. 154230050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87-48-9 | C8H4BrNO2 | 226.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-48-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.03
MDL Number MFCD00005719
InChI Key MBVCESWADCIXJN-UHFFFAOYSA-N
Synonym 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate
PubChem CID 6889
IUPAC Name 5-bromo-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)C(=O)N2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 251.0°C to 253.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 453
Formula Weight 226.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoisatin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 201-747-7

RUO – Research Use Only