Learn More
5-Bromoisatin, 90+%
CAS: 87-48-9 | C8H4BrNO2 | 226.029 g/mol
Supplier: Thermo Scientific Chemicals A1364136
Description
5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.
Solubility
Soluble in N,N dimethylformamide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 5-Bromoisatin | |
| ∼250°C (decomposition) | |
| MFCD00149345 | |
| 383760 | |
| Soluble in N,N dimethylformamide. | |
| C1=CC2=C(C=C1Br)C(=O)C(=O)N2 | |
| 226.029 | |
| 226.03 |
| 87-48-9 | |
| C8H4BrNO2 | |
| 500 g | |
| 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
| MBVCESWADCIXJN-UHFFFAOYSA-N | |
| 5-bromo-1H-indole-2,3-dione | |
| 6889 | |
| ≥90% |
Chemical Identifiers
| 87-48-9 | |
| 226.029 | |
| MBVCESWADCIXJN-UHFFFAOYSA-N | |
| 6889 | |
| C1=CC2=C(C=C1Br)C(=O)C(=O)N2 |
| C8H4BrNO2 | |
| MFCD00149345 | |
| 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
| 5-bromo-1H-indole-2,3-dione |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 201-747-7
RTECSNumber : NL7875000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only