missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. B47905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47905G 5 g
B47901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47905G Supplier TCI America Supplier No. B47905G

Chemical Identifiers

CAS 10406-06-1
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD05664007
InChI Key JVZMBSGNSAHFCY-UHFFFAOYSA-N
Synonym 5-bromoindole-3-carboxylic acid, 5-bromo-indole-3-carboxylic acid, 5-bromo-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid,5-bromo, 1h-indole-3-carboxylic acid, 5-bromo, 5-bromo-1h-indole-3-carboxylicacid, pubchem7982, acmc-1btmm, ksc173q4r, 5-bromo-3-indolecarboxylicacid
PubChem CID 7018243
IUPAC Name 5-bromo-1H-indole-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)C(=O)O

Specifications

Color White-Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 240.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-carboxylic Acid