CAS RN 10406-06-1
IUPAC Name:
5-bromo-1H-indole-3-carboxylic acid
Synonyms:
5-bromo-1H-indole-3-carboxylic acid
Molecular Weight (g/mol):
240.06
Molecular Formula:
C9H6BrNO2
InChi Key:
JVZMBSGNSAHFCY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CNC2=CC=C(Br)C=C12
1
–
1
of
1
results
1
5-Bromoindole-3-carboxylic Acid 98.0+%, TCI America™
CAS: 10406-06-1 Molecular Formula: C9H6BrNO2 Molecular Weight (g/mol): 240.056 MDL Number: MFCD05664007 InChI Key: JVZMBSGNSAHFCY-UHFFFAOYSA-N Synonym: 5-bromoindole-3-carboxylic acid,5-bromo-indole-3-carboxylic acid,5-bromo-3-indolecarboxylic acid,1h-indole-3-carboxylic acid,5-bromo,1h-indole-3-carboxylic acid, 5-bromo,5-bromo-1h-indole-3-carboxylicacid,pubchem7982,acmc-1btmm,ksc173q4r,5-bromo-3-indolecarboxylicacid PubChem CID: 7018243 IUPAC Name: 5-bromo-1H-indole-3-carboxylic acid SMILES: C1=CC2=C(C=C1Br)C(=CN2)C(=O)O