missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-m-xylene 98.0+%, TCI America™
$39.14 - $109.98
Chemical Identifiers
| CAS | 556-96-7 |
|---|---|
| Molecular Formula | C8H9Br |
| Molecular Weight (g/mol) | 185.064 |
| MDL Number | MFCD00000087 |
| InChI Key | LMFRTSBQRLSJHC-UHFFFAOYSA-N |
| Synonym | 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene |
| PubChem CID | 136357 |
| IUPAC Name | 1-bromo-3,5-dimethylbenzene |
| SMILES | CC1=CC(=CC(=C1)Br)C |
Chemical Identifiers
| 556-96-7 | |
| 185.064 | |
| LMFRTSBQRLSJHC-UHFFFAOYSA-N | |
| 136357 | |
| CC1=CC(=CC(=C1)Br)C |
| C8H9Br | |
| MFCD00000087 | |
| 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene | |
| 1-bromo-3,5-dimethylbenzene |
Specifications
| 556-96-7 | |
| 205°C | |
| Clear Liquid at 20°C | |
| MFCD00000087 | |
| LMFRTSBQRLSJHC-UHFFFAOYSA-N | |
| 1-bromo-3,5-dimethylbenzene | |
| 136357 | |
| ≥98.0% (GC) |
| Yellow | |
| 5 g | |
| C8H9Br | |
| 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene | |
| CC1=CC(=CC(=C1)Br)C | |
| 185.064 | |
| 185.06 | |
| 5-Bromo-m-xylene |
Safety and Handling
TSCA : No