missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-m-xylene, 98%

Catalog No. AAB2161022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2161022 100 g
AAB2161006 5 g
AAB2161014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2161022 Supplier Thermo Scientific Chemicals Supplier No. B2161022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 556-96-7 | C8H9Br | 185.064 g/mol

5-Bromo-m-xylene is used in the preparation of substituted 2,2'-bis(diphenylphosphanylmethyl)-1,1'-binaphthyl derivatives. It is also used to prepare o-tolyl-3,5-xylyl ether by reacting with o-cresol. Further, it is an organic building block used in the synthesis of various chemicals as well as in the preparation of tri-o-tolylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-m-xylene is used in the preparation of substituted 2,2′-bis(diphenylphosphanylmethyl)-1,1′-binaphthyl derivatives. It is also used to prepare o-tolyl-3,5-xylyl ether by reacting with o-cresol. Further, it is an organic building block used in the synthesis of various chemicals as well as in the preparation of tri-o-tolylphosphine.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 556-96-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000087
InChI Key LMFRTSBQRLSJHC-UHFFFAOYSA-N
Synonym 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene
PubChem CID 136357
IUPAC Name 1-bromo-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)Br)C

Specifications

Density 1.361
Quantity 100 g
Refractive Index 1.549
Beilstein 1926375
Boiling Point 204°C
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 185.07
Percent Purity 98%
Chemical Name or Material 5-Bromo-m-xylene
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only