missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-nitroindoline, 98%

Catalog No. p-7046970
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1699303 1 g
AAL1699306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1699303 Supplier Thermo Scientific Chemicals Supplier No. L1699303
Only null left

CAS: 80166-90-1 | C8H7BrN2O2 | 243.06 g/mol

5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 80166-90-1
Molecular Formula C8H7BrN2O2
Molecular Weight (g/mol) 243.06
MDL Number MFCD00005708
InChI Key VXKXMHDXFLFIFI-UHFFFAOYSA-N
Synonym 5-bromo-7-nitroindoline, 1h-indole, 5-bromo-2,3-dihydro-7-nitro, 5-bromo-7-nitro-1h indoline, 5-bromo-7-nitro-2,3-dihydroindole, 5-bromo-2,3-dihydro-7-nitro-1h-indole, pubchem7507, 5-bromo-7-nitro-indoline
PubChem CID 3018911
IUPAC Name 5-bromo-7-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC(Br)=CC2=C1NCC2

Specifications

Melting Point 128°C to 133°C
Quantity 1 g
UN Number UN2811
Beilstein 1373199
Solubility Information Insoluble in water.
Formula Weight 243.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-7-nitroindoline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 279-411-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only