accessibility menu, dialog, popup

UserName

ONOBrHN

CAS RN 80166-90-1

IUPAC Name: 5-bromo-7-nitro-2,3-dihydro-1H-indole
Synonyms: 5-bromo-7-nitro-2,3-dihydro-1H-indole
Molecular Weight (g/mol): 243.06
Molecular Formula: C8H7BrN2O2
InChi Key: VXKXMHDXFLFIFI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CC2=C1NCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results