missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-7-nitroindoline, 98%
Description
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 128°C to 133°C |
| Quantity | 1 g |
| UN Number | UN2811 |
| Beilstein | 1373199 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 243.06 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Bromo-7-nitroindoline |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 279-411-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only