missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-6-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt 98.0+%, TCI America™
$58.85 - $351.25
Chemical Identifiers
| CAS | 144110-43-0 |
|---|---|
| Molecular Formula | C20H26BrClN2O7 |
| Molecular Weight (g/mol) | 521.789 |
| MDL Number | MFCD00153929 |
| InChI Key | HGZDFBMYVMPFHR-CWBGJGNPSA-N |
| Synonym | 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt |
| PubChem CID | 134129378 |
| IUPAC Name | (2S,3R,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine |
| SMILES | C1CCC(CC1)N.C1=C2C(=CC(=C1Br)Cl)NC=C2OC3C(C(C(C(O3)C(=O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
B412810MG
|
N/A | 10 mg |
Each for $58.85
|
|
|||||
|
B4128100MG
|
N/A | 100 mg |
Each for $351.25
|
|
|||||
Chemical Identifiers
| 144110-43-0 | |
| 521.789 | |
| HGZDFBMYVMPFHR-CWBGJGNPSA-N | |
| 134129378 | |
| C1CCC(CC1)N.C1=C2C(=CC(=C1Br)Cl)NC=C2OC3C(C(C(C(O3)C(=O)O)O)O)O |
| C20H26BrClN2O7 | |
| MFCD00153929 | |
| 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt | |
| (2S,3R,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine |
Specifications
| 144110-43-0 | |
| C20H26BrClN2O7 | |
| 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt | |
| C1CCC(CC1)N.C1=C2C(=CC(=C1Br)Cl)NC=C2OC3C(C(C(C(O3)C(=O)O)O)O)O | |
| 521.789 | |
| 521.79 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00153929 | |
| HGZDFBMYVMPFHR-CWBGJGNPSA-N | |
| (2S,3R,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine | |
| 134129378 | |
| ≥98.0% (HPLC) | |
| 5-Bromo-6-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt [for Biochemical Research] |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)