missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt 98.0+%, TCI America™

Catalog No. B412810MG
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B412810MG 10 mg
B4128100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B412810MG Supplier TCI America Supplier No. B412810MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144110-43-0
Molecular Formula C20H26BrClN2O7
Molecular Weight (g/mol) 521.789
MDL Number MFCD00153929
InChI Key HGZDFBMYVMPFHR-CWBGJGNPSA-N
Synonym 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt
PubChem CID 134129378
IUPAC Name (2S,3R,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine
SMILES C1CCC(CC1)N.C1=C2C(=CC(=C1Br)Cl)NC=C2OC3C(C(C(C(O3)C(=O)O)O)O)O

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 521.79
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-6-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt [for Biochemical Research]
TSCA No
Recommended Storage Freezer (-20 °C)