missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-6-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt 98.0+%, TCI America™
Supplier: TCI America B4128100MG
Specifications
| 5-Bromo-6-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt [for Biochemical Research] | |
| White-Yellow | |
| MFCD00153929 | |
| 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt | |
| C1CCC(CC1)N.C1=C2C(=CC(=C1Br)Cl)NC=C2OC3C(C(C(C(O3)C(=O)O)O)O)O | |
| 521.789 | |
| 521.79 | |
| Crystalline Powder |
| 144110-43-0 | |
| C20H26BrClN2O7 | |
| 100 mg | |
| HGZDFBMYVMPFHR-CWBGJGNPSA-N | |
| (2S,3R,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine | |
| 134129378 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 144110-43-0 | |
| 521.789 | |
| HGZDFBMYVMPFHR-CWBGJGNPSA-N | |
| 134129378 | |
| C1CCC(CC1)N.C1=C2C(=CC(=C1Br)Cl)NC=C2OC3C(C(C(C(O3)C(=O)O)O)O)O |
| C20H26BrClN2O7 | |
| MFCD00153929 | |
| 5-Bromo-6-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, Magenta-Gluc CHA Salt, Magenta-GlcA CHA Salt | |
| (2S,3R,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)