missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-5-nitro-1,3-dioxane 98.0+%, TCI America™

Catalog No. B37695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37695G Supplier TCI America Supplier No. B37695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30007-47-7
Molecular Formula C4H6BrNO4
Molecular Weight (g/mol) 211.999
MDL Number MFCD00101855
InChI Key XVBRCOKDZVQYAY-UHFFFAOYSA-N
Synonym bronidox, 1,3-dioxane, 5-bromo-5-nitro, 5-bromo-5-nitro-m-dioxane, unii-u184i9qbnm, 5-brom-5-nitro-1,3-dioxan, m-dioxane, 5-bromo-5-nitro, 5-brom-5-nitro-1,3-dioxan german, u184i9qbnm, bronidox l, acmc-209hcf
PubChem CID 1807
IUPAC Name 5-bromo-5-nitro-1,3-dioxane
SMILES C1C(COCO1)([N+](=O)[O-])Br

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-5-nitro-1,3-dioxane [for Biochemical Research]
RTECSNumber JG9650000
TSCA Yes
Recommended Storage Refrigerator